missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RA/IL-17R Antibody (49M4D2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25271AF488
This item is not returnable.
View return policy
Description
IL-17RA/IL-17R Monoclonal specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-17RA/IL-17R | |
| Monoclonal | |
| Alexa Fluor 488 | |
| CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 23765 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 49M4D2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| IL17RA | |
| Within the range of amino acids 186-267 of human IL-17RA were used as the immunogen for the antibody. | |
| 0.1 mL | |
| Primary | |
| This product reacts with Human. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction