missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RA/IL-17R Antibody (657603), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4481M
This item is not returnable.
View return policy
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Mouse samples. It is validated for Flow Cytometry, Neutralization, CyTOF
Specifications
| IL-17RA/IL-17R | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Neutralization, CyTOF | |
| 657603 | |
| Flow Cytometry, Neutralization, CyTOF-ready | |
| CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RA/IL-17 R, Ser32-Trp322, Accession # Q60943 | |
| 0.1 mL | |
| Innate Immunity | |
| 23765 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction