missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF8/CD316 Antibody (8A12), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21498H
This item is not returnable.
View return policy
Description
IGSF8/CD316 Monoclonal antibody specifically detects IGSF8/CD316 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| IGSF8/CD316 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 8A12 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD316, CD316 antigen, CD81P3LIR-D1, EWI-2, EWI2CD81 partner 3, Glu-Trp-Ile EWI motif-containing protein 2, IgSF8, immunoglobulin superfamily, member 8, KCT4, KCT-4, Keratinocytes-associated transmembrane protein 4, PGRLimmunoglobulin superfamily member 8 | |
| Human leukocytes | |
| 0.1 mL | |
| Neuroscience | |
| 93185 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction