missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RA/IL-17R Antibody (SR1226), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | IL-17RA/IL-17R |
|---|---|
| Clone | SR1226 |
| Dilution | Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668414
|
Novus Biologicals
NBP3-22223-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603964
|
Novus Biologicals
NBP3-22223-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| IL-17RA/IL-17R | |
| Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
| A synthesized peptide derived from human IL-17RA/IL-17R (Uniprot #: Q96F46) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1226 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Innate Immunity | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 23765 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title