missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RC Antibody (209), Alexa Fluor™ 405 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89976AF405
This item is not returnable.
View return policy
Description
IL-17RC Monoclonal antibody specifically detects IL-17RC in Human samples. It is validated for ELISA
Specifications
| IL-17RC | |
| Monoclonal | |
| Alexa Fluor 405 | |
| FLJ95963, FLJ96005, IL-17 receptor C, IL17F receptor, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-17RC (NP_116121.2; Met1-Ala454). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 209 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 84818 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction