missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RE Antibody (934832) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8358F
This item is not returnable.
View return policy
Description
IL-17RE Monoclonal antibody specifically detects IL-17RE in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Specifications
| IL-17RE | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 132014 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 934832 | |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| IL-17 receptor E, interleukin 17 receptor E | |
| Chinese hamster ovary cell line CHO-derived recombinant human IL-17 RE, Met1-His454, Accession # Q8NFR9 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction