missing translation for 'onlineSavingsMsg'
Learn More

IL-17RE Antibody (944904) [Alexa Fluor™ 532], Novus Biologicals™

Product Code. 18549867 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18549867 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18549867 Supplier Novus Biologicals Supplier No. FAB7997X

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-17RE Monoclonal antibody specifically detects IL-17RE in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-17RE
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 944904
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias IL-17 receptor E, interleukin 17 receptor E
Host Species Rat
Immunogen Mouse myeloma cell line NSO-derived recombinant mouse IL-17 RE, Met1-His414, Accession # Q8BH06
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132014
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.