missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RE Antibody (944904) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7997JF585
This item is not returnable.
View return policy
Description
IL-17RE Monoclonal antibody specifically detects IL-17RE in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| IL-17RE | |
| Monoclonal | |
| Janelia Fluor 585 | |
| IL-17 receptor E, interleukin 17 receptor E | |
| Mouse myeloma cell line NSO-derived recombinant mouse IL-17 RE, Met1-His414, Accession # Q8BH06 | |
| RUO | |
| 132014 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 944904 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction