missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18 BPa/IL18BP Antibody (224), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89445CL1
This item is not returnable.
View return policy
Description
IL-18 BPa/IL18BP Monoclonal antibody specifically detects IL-18 BPa/IL18BP in Human samples. It is validated for ELISA
Specifications
| IL-18 BPa/IL18BP | |
| Monoclonal | |
| CoraFluor 1 | |
| IL-18BP, IL18BPa, interleukin 18 binding protein, interleukin-18-binding protein, MC51L-53L-54L homolog gene product, tadekinig-alfa | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-18 BPa/IL18BP (Accession#: NP_001034748.1; Met1-Gly194). | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 10068 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 224 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction