missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18 BPa/IL18BP Antibody (MM0379-10G36), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11678DL594
This item is not returnable.
View return policy
Description
IL-18 BPa/IL18BP Monoclonal antibody specifically detects IL-18 BPa/IL18BP in Human samples. It is validated for Western Blot, Neutralization
Specifications
| IL-18 BPa/IL18BP | |
| Monoclonal | |
| DyLight 594 | |
| IL-18BP, IL18BPa, interleukin 18 binding protein, interleukin-18-binding protein, MC51L-53L-54L homolog gene product, tadekinig-alfa | |
| Human recombinant IL-18 BPa/IL18BP | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 10068 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Neutralization | |
| MM0379-10G36 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction