missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18/IL-1F4 Antibody (74801), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB646W
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Monoclonal antibody specifically detects IL-18/IL-1F4 in Human samples. It is validated for Western Blot, Flow Cytometry, KnockDown
Specifications
| IL-18/IL-1F4 | |
| Monoclonal | |
| DyLight 650 | |
| Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
| E. coli-derived recombinant human IL-18/IL-1F4, Accession # Q14116 | |
| 0.5 mL | |
| Angiogenesis, Apoptosis, Cancer, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Endocrinology, Immunology, Innate Immunity | |
| 3606 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, KnockDown | |
| 74801 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction