missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18/IL-1F4 Antibody (925008), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) FAB2548MFV610
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Monoclonal antibody specifically detects IL-18/IL-1F4 in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| IL-18/IL-1F4 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Endocrinology, Immunology, Innate Immunity | |
| 3606 | |
| Store at 4C in the dark. |
| ELISA, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 925008 | |
| ELISA, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
| E. coli-derived recombinant Rhesus monkey IL-18/IL-1F4, Tyr37-Asp193, Accession # Q14116 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction