missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-18/IL-1F4 Antibody (ABT-325), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28597-1mg
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Monoclonal antibody specifically detects IL-18/IL-1F4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-18/IL-1F4 | |
| Monoclonal | |
| Unconjugated | |
| Q14116 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| ABT-325 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
| IL-18 | |
| 1 mg | |
| Angiogenesis, Apoptosis, Cancer, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Endocrinology, Immunology, Innate Immunity | |
| 3606 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction