Learn More
Invitrogen™ IL-18 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA547344
Product Code. 15587832
  • € 514.00 / 100µg
Description
This antibody detects rhesus macaque IL-18/IL-1F4 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse IL-18, recombinant rat IL-18, and recombinant porcine IL-18 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-18
Polyclonal
Unconjugated
IL18
cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320
Goat
Antigen affinity chromatography
RUO
3606, 574151
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Neutralization, Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 5% trehalose and No Preservative
Q14116
IL18
E. coli-derived recombinant rhesus macaque IL-18/IL-1F4 Tyr37-Asp193.
100 μg
Primary
Human, Rhesus Macaque