missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18R alpha/IL-1 R5 Antibody (112614) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1216JF585
This item is not returnable.
View return policy
Description
IL-18R alpha/IL-1 R5 Monoclonal antibody specifically detects IL-18R alpha/IL-1 R5 in Mouse samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| IL-18R alpha/IL-1 R5 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD218a;CDw218a;IL18R1;IL18RA;IL-18Ralpha;IL-18R-alpha;IL18Ralpha2;IL1RRP;IL-1Rrp;Interleukin-18 receptor 1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha /IL-1 R5, Ser19-Gly326, Accession # Q61098 | |
| 0.5 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Western Blot, CyTOF | |
| 112614 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 8809 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction