Learn More
IL-18R alpha/IL-1 R5 Antibody (70625), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB840MFV610
Product Code. 18508989
  • € 708.00 / 0.10mL
Description
IL-18R alpha/IL-1 R5 Monoclonal antibody specifically detects IL-18R alpha/IL-1 R5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-18R alpha/IL-1 R5
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
0.1 mL
Primary
Human
IgG1
Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF
70625
Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-ready
CD218a;CDw218a;IL18R1;IL18RA;IL-18Ralpha;IL-18R-alpha;IL18Ralpha2;IL1RRP;IL-1Rrp;Interleukin-18 receptor 1
Mouse myeloma cell line NS0-derived recombinant human IL-18 R alpha/IL-1 R5, Glu20-Arg329, Accession # Q13478
RUO
8809
Store at 4C in the dark.