missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-19 Antibody (152112), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1035L
This item is not returnable.
View return policy
Description
IL-19 Monoclonal antibody specifically detects IL-19 in Human samples. It is validated for Flow Cytometry
Specifications
| IL-19 | |
| Monoclonal | |
| DyLight 550 | |
| IL-10C, IL-19, interleukin 19, MDA1, melanoma differentiation associated protein-like protein, NG.1Melanoma differentiation-associated protein-like protein, ZMDA1interleukin-19 | |
| E. coli-derived recombinant human IL-19, Leu25-Ala177, Accession # AAN40906 | |
| 0.5 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 29949 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 152112 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction