missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-19 Antibody (350105) [Janelia Fluor™ 669], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB29151JF669
This item is not returnable.
View return policy
Description
IL-19 Monoclonal antibody specifically detects IL-19 in Mouse samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| IL-19 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| IL-10C, IL-19, interleukin 19, MDA1, melanoma differentiation associated protein-like protein, NG.1Melanoma differentiation-associated protein-like protein, ZMDA1interleukin-19 | |
| E. coli-derived recombinant mouse IL-19, Leu25-Ala176, Accession # Q8CJ70 | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 29949 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 350105 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction