Learn More
IL-1F6 / IL-36 alpha Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24173-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by IL1F6 is a cytokine that can activate NF-kappa-B and MAPK signaling pathways to generate an inflammatory response. The encoded protein functions primarily in skin and demonstrates increased expression in psoriasis. In addition, decreased expression of this gene has been linked to a poor prognosis in both hepatocellular carcinoma and colorectal cancer patients.Specifications
| IL-1F6 / IL-36 alpha | |
| Polyclonal | |
| Unconjugated | |
| IL36A | |
| FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL 1 epsilon, IL 1F6, IL1(EPSILON), IL1E, IL1F6, Interleukin 1 epsilon, Interleukin 1 family member 6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27179 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UHA7 | |
| IL36A | |
| IL-1F6/IL-36alpha Fusion Protein Ag21487 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |