missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1ra/IL-1F3/IL1RN Antibody (OTI2B1), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71022R
This item is not returnable.
View return policy
Description
IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| IL-1ra/IL-1F3/IL1RN | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IL1RN | |
| Full length recombinant protein of human IL-1ra/IL-1F3/IL1RN (NP_776215) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI2B1 | |
| Western Blot, Immunohistochemistry | |
| DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
| Mouse | |
| Affinity Purified | |
| Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3557.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction