missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-1RAcP Polyclonal Antibody
GREENER_CHOICE

Product Code. p-200128803
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17790624 200 μL 200µL
17713284 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17790624 Supplier Invitrogen™ Supplier No. PA5119222

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IL-1RAcP Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a component of the interleukin 1 receptor complex, which initiates signaling events that result in the activation of interleukin 1-responsive genes. Alternative splicing of this gene results in membrane-bound and soluble isoforms differing in their C-terminus. The ratio of soluble to membrane-bound forms increases during acute-phase induction or stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1RAcP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene IL1RAP
Gene Accession No. Q9NPH3
Gene Alias 6430709H04Rik; AI255955; AV239853; C3orf13; hide; IL 1R 3; IL 1R3; IL 1RAcP; IL 1RAP; IL-1 receptor accessory protein; IL1R3; IL-1R3; IL-1R-3; IL1RAcP; IL-1RAcp; Il1racpb; Il1rap; ILN; Interleukin; interleukin 1 receptor accessory protein; interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta; Interleukin-33 receptot beta chain
Gene Symbols IL1RAP
Host Species Rabbit
Immunogen Recombinant protein Interleukin 1 Receptor Accessory Protein. The antigen corresponds to amino acid range 65-249 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3556
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.