missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-1RAcP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580723
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IL-1RAcP/IL-1R3.
This gene encodes a component of the interleukin 1 receptor complex, which initiates signaling events that result in the activation of interleukin 1-responsive genes. Alternative splicing of this gene results in membrane-bound and soluble isoforms differing in their C-terminus. The ratio of soluble to membrane-bound forms increases during acute-phase induction or stress.
Specifications
| IL-1RAcP | |
| Polyclonal | |
| Unconjugated | |
| IL1RAP | |
| 6430709H04Rik; AI255955; AV239853; C3orf13; hide; IL 1R 3; IL 1R3; IL 1RAcP; IL 1RAP; IL-1 receptor accessory protein; IL1R3; IL-1R3; IL-1R-3; IL1RAcP; IL-1RAcp; Il1racpb; Il1rap; ILN; Interleukin; interleukin 1 receptor accessory protein; interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta; Interleukin-33 receptot beta chain | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 3556 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1.0 mg/mL | |
| PBS with no preservative | |
| Q9NPH3 | |
| IL1RAP | |
| Recombinant Human IL-1RAcP/IL-1R3 protein (Met1-Glu359). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction