missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1RL1/ST2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | IL-1RL1/ST2 |
|---|---|
| Concentration | 650 μg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233708
|
Proteintech
30194-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236939
|
Proteintech
30194-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IL-1RL1/ST2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IL-1RL1/ST2. This antibody recognizes human antigen. The IL-1RL1/ST2 antibody has been validated for the following applications: WB, FC, ELISA analysis.Specifications
| IL-1RL1/ST2 | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DER4; FIT-1; MGC32623; ST2; ST2L; ST2V; T1 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 650 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 9173 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title