missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1Rrp2/IL-1R6 Antibody (imsidolimab) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28330JF669
This item is not returnable.
View return policy
Description
IL-1Rrp2/IL-1R6 Monoclonal antibody specifically detects IL-1Rrp2/IL-1R6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-1Rrp2/IL-1R6 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2 | |
| IL-1RL2/ IL-36R | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| imsidolimab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 8808 | |
| Store at 4°C in the dark. | |
| IgG4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction