missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 Antibody (011), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90002AF594
This item is not returnable.
View return policy
Description
IL-2 Monoclonal specifically detects IL-2 in Human samples. It is validated for ELISA.
Specifications
| IL-2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 11 | |
| ELISA | |
| aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-2 (P60568-1; Ala21-Thr153). | |
| 0.1 mL | |
| Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers | |
| 3558 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction