missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 Antibody (1019308), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB103561G
This item is not returnable.
View return policy
Description
IL-2 Monoclonal antibody specifically detects IL-2 in Human samples. It is validated for Flow Cytometry
Specifications
| IL-2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1019308 | |
| Intracellular Staining by Flow Cytometry | |
| aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
| E. coli -derived human IL-2, Ala21-Thr153, Accession # P60568 | |
| 0.1 mL | |
| Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers | |
| 3558 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur