Learn More
IL-2 Antibody (5334R), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB202RMFV450
Product Code. 30061025
  • € 674.00 / 0.10mL
Description
IL-2 Monoclonal antibody specifically detects IL-2 in Human samples. It is validated for Neutralization, Flow Cytometry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-2
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
Neutralization, Flow Cytometry, Immunocytochemistry
5334R
Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry
aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
E. coli-derived recombinant human IL-2, Ala21-Thr153, Accession # NP_000577
0.1 mL
Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers
3558
Store at 4C in the dark.