missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 Antibody (5343), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10442V
This item is not returnable.
View return policy
Description
IL-2 Monoclonal antibody specifically detects IL-2 in Human samples. It is validated for Neutralization, Flow Cytometry, CyTOF
Specifications
| IL-2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Neutralization, Flow Cytometry, CyTOF | |
| 5343 | |
| Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
| E. coli-derived recombinant human IL-2, Ala21-Thr153, Accession # NP_000577 | |
| 0.1 mL | |
| Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers | |
| 3558 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction