missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
IL-2 Antibody (B-G5) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14593JF525
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IL-2 Monoclonal antibody specifically detects IL-2 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Functional Assay,Enzyme-linked Immunosorbent Spot
Tekniske data
| IL-2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
| Natural human IL-2 | |
| 0.1 mL | |
| Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers | |
| 3558 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Western Blot, Functional Assay, ELISpot | |
| B-G5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion