Learn More
IL-2 Mouse anti-Human, CoraLite 488, Clone: 2F11C5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-60306-100UL
Product Code. 17472543
  • € 450.00 / 100µL
Description
The IL-2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-2. This antibody recognizes human antigen. The IL-2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
IL-2
Monoclonal
CoraLite 488
100 μL
3558
IgG2a
Immunofluorescence
2F11C5