missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-2 R beta Antibody (03) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30744MFV500
This item is not returnable.
View return policy
Description
IL-2 R beta Monoclonal antibody specifically detects IL-2 R beta in Human samples. It is validated for ELISA
Specifications
| IL-2 R beta | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
| Produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-2 R beta (RefSeq#: NP_000869.1; Met1-Asp239) | |
| 0.1 mL | |
| Immunology | |
| 3560 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA | |
| 03 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction