missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 R beta Antibody (27302), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB224PEATT594
This item is not returnable.
View return policy
Description
IL-2 R beta Monoclonal antibody specifically detects IL-2 R beta in Human samples. It is validated for Flow Cytometry
Specifications
| IL-2 R beta | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 27302 | |
| Flow Cytometry | |
| CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-2 R beta , Ala27-Asp239, Accession # NP_000869 | |
| 0.1 mL | |
| Immunology | |
| 3560 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction