Learn More
IL-2 Rat, PE, Clone: JES6-5H4, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB80-PE-100ug
 View more versions of this product
Product Code. 15912538
  • € 301.35 / 100µg
Specifications
IL-2
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Il2
Protein A/G Purified
RUO
16183
IgG2b κ
Flow Cytometry
JES6-5H4
PE
NP_032392
Rat
100 μg
Primary
Store at 2-4°C.