missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-20 Antibody (03) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26912AF700
This item is not returnable.
View return policy
Description
IL-20 Monoclonal antibody specifically detects IL-20 in Human samples. It is validated for ELISA,Sandwich ELISA Detection
Specifications
| IL-20 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Cytokine Zcyto10, IL10D, IL-20ZCYTO10four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-20 (Uniprot#: Q9NYY1; Leu25-Glu176). | |
| 0.1 mL | |
| Biologically Active Proteins, Signal Transduction | |
| 50604 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Sandwich ELISA Detection | |
| 03 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction