missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-20 Antibody (044), DyLight 488 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90162G
This item is not returnable.
View return policy
Description
IL-20 Monoclonal antibody specifically detects IL-20 in Human samples. It is validated for ELISA, ELISA
Specifications
| IL-20 | |
| Monoclonal | |
| DyLight 488 | |
| Cytokine Zcyto10, IL10D, IL-20ZCYTO10four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-20 (Q9NYY1; Leu25-Glu176). | |
| 0.1 mL | |
| Biologically Active Proteins, Signal Transduction | |
| 50604 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 044 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction