Learn More
Novus Biologicals™ IL-20 Antibody (Cheng Kung U. patent anti-IL-20) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28741-50ug
 View more versions of this product
Product Code. 30048610
  • € 213.00 / 50µg
Description
IL-20 Monoclonal antibody specifically detects IL-20 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-20
Monoclonal
Unconjugated
Q9NYY1
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Cheng Kung U. patent anti-IL-20
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Cytokine Zcyto10, IL10D, IL-20ZCYTO10four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907
IL-20
50 μg
Biologically Active Proteins, Signal Transduction
50604