missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-20 R beta/FNDC6 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98428G
This item is not returnable.
View return policy
Description
IL-20 R beta/FNDC6 Polyclonal specifically detects IL-20 R beta/FNDC6 in Rat samples. It is validated for Western Blot, ELISA.
Specifications
| IL-20 R beta/FNDC6 | |
| Polyclonal | |
| Western Blot, ELISA | |
| FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923 | |
| Produced in rabbits immunized with purified, recombinant IL-20 R beta/FNDC6 (B8K1X9; Met1-Pro227). | |
| 0.1 mL | |
| Primary | |
| Rat | |
| Purified |
| Western Blot, ELISA | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 53833 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction