missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-20R alpha Antibody (173714) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB11762JF525
This item is not returnable.
View return policy
Description
IL-20R alpha Monoclonal antibody specifically detects IL-20R alpha in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| IL-20R alpha | |
| Monoclonal | |
| Janelia Fluor 525 | |
| class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-20 R alpha , Val30-Lys250, Accession # Q9UHF4 | |
| 0.5 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Western Blot, CyTOF | |
| 173714 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 53832 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction