missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-21 Antibody (149204), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB594T
This item is not returnable.
View return policy
Description
IL-21 Monoclonal antibody specifically detects IL-21 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| IL-21 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 149204 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform | |
| E. coli-derived recombinant mouse IL-21, Pro25-Ser146, Accession # Q9ES17 | |
| 0.5 mL | |
| Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction | |
| 59067 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction