missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-21 Antibody (14k5H3), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27336AF488
This item is not returnable.
View return policy
Description
IL-21 Monoclonal specifically detects IL-21 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| IL-21 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Q9HBE4 | |
| IL21 | |
| Full length recombinant protein | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction | |
| 59067 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 14k5H3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction