missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-21 Antibody (14k5H3), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27336AF532
This item is not returnable.
View return policy
Description
IL-21 Monoclonal specifically detects IL-21 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| IL-21 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| IL21 | |
| Full length recombinant protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 14k5H3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform | |
| Mouse | |
| Protein G purified | |
| Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction | |
| 59067.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction