missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-21 Antibody (227), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26073C
This item is not returnable.
View return policy
Description
IL-21 Monoclonal antibody specifically detects IL-21 in Mouse samples. It is validated for ELISA
Specifications
| IL-21 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 227 | |
| ELISA | |
| IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform | |
| Rabbit immunized with purified, recombinant Mouse IL-21 (Uniprot#: Q9ES17, Pro25-Ser146) | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction | |
| 59067 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction