missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-21R Antibody (152512), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9911D
This item is not returnable.
View return policy
Description
IL-21R Monoclonal antibody specifically detects IL-21R in Human samples. It is validated for Flow Cytometry
Specifications
| IL-21R | |
| Monoclonal | |
| DyLight 350 | |
| CD360 antigen, IL-21 receptor, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-21 R, Cys20-Pro236, Accession # Q9HBE5 | |
| 0.5 mL | |
| Chemokines and Cytokines, Cytokine Research, Immunology | |
| 50615 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 152512 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction