missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-22 Antibody (140301), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB582G
This item is not returnable.
View return policy
Description
IL-22 Monoclonal antibody specifically detects IL-22 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| IL-22 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 140301 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| E. coli-derived recombinant mouse IL-22, Leu34-Val179, Accession # Q9JJY9 | |
| 0.5 mL | |
| Biologically Active Proteins, Cytokine Research, Innate Immunity | |
| 50616 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction