missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-22 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41245AF647
This item is not returnable.
View return policy
Description
IL-22 Polyclonal antibody specifically detects IL-22 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| IL-22 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 647 | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| Antibody was raised against a 17 amino acid peptide near the center of human IL-22 . The immunogen is located within amino acids 90 - 140 of IL-22. Amino Acid Squence: EEVLFPQSDRFQPYMQE | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research, Innate Immunity | |
| 50616 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction