missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-22BP/IL22 RA2 Antibody (MM0397-6G47), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11700AF700
This item is not returnable.
View return policy
Description
IL-22BP/IL22 RA2 Monoclonal specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for Western Blot.
Specifications
| IL-22BP/IL22 RA2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IL22RA2 | |
| Human recombinant IL-22BP/IL22 RA2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0397-6G47 | |
| Western Blot | |
| class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 receptor subunit alpha-2, IL-22BPCRF2X, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16 | |
| Mouse | |
| Protein G purified | |
| Cytokine Research, Immunology, Signal Transduction | |
| 116379 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction