missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-22R alpha 1 Antibody (496514), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB42941L
This item is not returnable.
View return policy
Description
IL-22R alpha 1 Monoclonal antibody specifically detects IL-22R alpha 1 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| IL-22R alpha 1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 496514 | |
| Flow Cytometry, CyTOF-ready | |
| CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R1, IL-22RA1, IL-22R-alpha-1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-22 R alpha 1, Thr18-Ala228, Accession # Q80XZ4 | |
| 0.1 mL | |
| Cytokine Research, Immunology, Signal Transduction | |
| 58985 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction