Learn More
IL-22R alpha 1 Antibody (496514), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB42941Y
Product Code. 18548359
  • ONLINE EXCLUSIVE PRICE
    € 706.00
    € 667.80 / 0.10mL
    Save € 38.20
    5% Off
Description
IL-22R alpha 1 Monoclonal antibody specifically detects IL-22R alpha 1 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-22R alpha 1
Monoclonal
DyLight 680
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry, CyTOF
496514
Flow Cytometry, CyTOF-ready
CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R1, IL-22RA1, IL-22R-alpha-1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11
Mouse myeloma cell line NS0-derived recombinant mouse IL-22 R alpha 1, Thr18-Ala228, Accession # Q80XZ4
0.1 mL
Cytokine Research, Immunology, Signal Transduction
58985
Store at 4°C in the dark.
IgG2a