missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-23 Antibody (mirikizumab) - Humanized, IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28749-100ug
This item is not returnable.
View return policy
Description
IL-23 Monoclonal antibody specifically detects IL-23 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-23 | |
| Monoclonal | |
| Unconjugated | |
| Q9NPF7 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| mirikizumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12B, IL-23, IL-23 subunit alpha, IL-23A, IL-23-A, IL-23p19, IL23P19P19, interleukin 12, p40, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-12 beta chain, interleukin-12 subunit beta, Interleukin-23 subunit p19, interleukin-six, G-CSF related factor, JKA3 induced upon T-cell activation, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, P19, SGRF | |
| IL-23 | |
| 100 μg | |
| Cancer, Cytokine Research, Innate Immunity | |
| 51561 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction