Learn More
IL-23A/IL-23 P19 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-77257F
Product Code. 18445910
  • € 630.00 / 0.10mL
Description
IL-23A/IL-23 P19 Polyclonal antibody specifically detects IL-23A/IL-23 P19 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-23A/IL-23 P19
Polyclonal
PBS
Rabbit
Peptide affinity purified
RUO
Primary
Human, Mouse
Purified
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
FITC
IL-23, IL-23A, IL-23-A, IL-23p19, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRFIL-23 subunit alpha
Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human IL-23. The immunogen is located within the last 50 amino acids of IL-23. Amino Acid Squence: QIPSLSPSQPWQRL
0.1 mL
Cancer, Cytokine Research, Innate Immunity
51561
Store at 4°C in the dark.